Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010892.3 | NP_035022.2 | 2 | 458 | ||
GenBank mRNA | AF007247.1 | - | 2 | 372 | ||
AF013166.1 | - | 2 | 342 | |||
AK077627.1 | - | 2 | 422 | |||
AK147072.1 | - | 2 | 434 | |||
AK164467.1 | - | 2 | 401 | |||
AK169330.1 | - | 3 | 349 | |||
AK170179.1 | - | 2 | 378 | |||
AK171277.1 | - | 2 | 378 | |||
AK171387.1 | - | 2 | 378 | |||
BC010302.1 | - | 2 | 380 | 3597227 | ||
BC011316.1 | - | 2 | 378 | 3674001 | ||
BC057576.1 | - | 2 | 398 | 6810914 | ||
CF738658.1 | - | 2 | 485 | |||
U95610.1 | - | 2 | 369 |