Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113213.1 | NP_001106684.1 | 10 | 1251 | ||
NM_001113214.1 | NP_001106685.1 | 10 | 1251 | |||
NM_001177659.1 | NP_001171130.1 | 10 | 1254 | |||
NM_001177661.1 | NP_001171132.1 | 9 | 1097 | |||
NM_013615.3 | NP_038643.1 | 10 | 1224 | |||
GenBank mRNA | AF000968.1 | - | 8 | 1071 | ||
AF034105.1 | - | 10 | 1187 | |||
AK143726.1 | - | 10 | 1158 | |||
AK148523.1 | - | 10 | 1164 | |||
AK165588.1 | - | 10 | 1155 | |||
AK168415.1 | - | 10 | 1153 | |||
BC057001.1 | - | 11 | 1308 | 6827260 |