Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OPRM1 SIRNA Assays ›
          Gene Symbol:
          Oprm1
          Gene Name:
          opioid receptor, mu 1
          Gene Aliases:
          M-OR-1, MOP-R, MOR-1, MOR-1O, Oprm, mor, muOR
          Chromosome Location:
          Chr.10: 6758593 - 7038209 on Build GRCm38
          UniGene ID:
          Mm.438000
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS207314
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Oprm1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039652.2 NP_001034741.1 2 728
          GenBank mRNA AB047546.1 - 2 643
          AB441734.1 - 2 429
          AB441735.1 - 2 429
          AB441736.1 - 2 429
          AB441737.1 - 2 429
          AB441738.1 - 2 429
          AB441739.1 - 2 429
          AB441740.1 - 2 429
          AB441741.1 - 2 429
          AB441742.1 - 2 429
          AB441743.1 - 2 429
          AF062753.1 - 2 495
          AF062755.1 - 2 330
          AF074972.1 - 2 330
          AF074973.1 - 2 495
          AF074974.1 - 2 495
          AF167565.1 - 2 495
          AF167566.1 - 2 495
          AF167567.1 - 2 495
          AF167568.1 - 2 495
          AF260306.1 - 3 658
          AF260307.1 - 3 832
          AF260308.1 - 4 787
          AF260309.1 - 3 392
          AF260310.1 - 3 351
          AF260311.1 - 2 243
          AF346812.1 - 2 495
          AF346813.1 - 2 495
          AF346814.1 - 2 495
          AF400246.1 - 3 658
          AF400247.1 - 3 832
          AF400248.1 - 4 787
          AK038389.1 - 2 469
          AY036621.1 - 2 495
          AY160190.1 - 2 495
          AY923184.2 - 4 761
          BC119545.1 - 2 495 40056952
          DQ363376.1 - 2 329
          DQ363377.1 - 2 329
          DQ868787.1 - 2 495
          DQ868788.2 - 2 495
          EF105311.1 - 2 495
          EF105312.1 - 2 495
          EF105313.1 - 2 495
          EF105314.1 - 2 495
          U19380.1 - 2 684
          U26915.1 - 2 711

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          chemical synaptic transmission
          locomotory behavior
          sensory perception of pain
          negative regulation of cAMP biosynthetic process
          adenylate cyclase-inhibiting opioid receptor signaling pathway
          positive regulation of appetite
          cellular response to stress
          opioid receptor signaling pathway
          eating behavior
          positive regulation of cAMP-mediated signaling
          negative regulation of cAMP-mediated signaling
          negative regulation of nitric oxide biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          response to ethanol
          behavioral response to ethanol
          positive regulation of neurogenesis
          negative regulation of cytosolic calcium ion concentration
          regulation of sensory perception of pain
          excitatory postsynaptic potential
          negative regulation of Wnt protein secretion
          positive regulation of ERK1 and ERK2 cascade
          calcium ion transmembrane transport
          cellular response to morphine
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          G-protein alpha-subunit binding
          signal transducer activity
          G-protein coupled receptor activity
          beta-endorphin receptor activity
          opioid receptor activity
          voltage-gated calcium channel activity
          protein binding
          protein C-terminus binding
          protein domain specific binding
          filamin binding
          G-protein beta-subunit binding
          morphine receptor activity
          neuropeptide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gl4ql:80/100.66.76.174:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0