siRNA Details
Gene Symbol: Oca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021879.2 | NP_068679.1 | 11 | 1254 | ||
GenBank mRNA | AK051897.1 | - | 10 | 1095 | ||
AK087494.1 | - | 12 | 1514 | |||
BC119220.1 | - | 11 | 1224 | 8733969 | ||
BC120549.1 | - | 11 | 1224 | 40129472 | ||
M97900.1 | - | 11 | 1254 |
