Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDGFRB SIRNA Assays ›
          Gene Symbol:
          Pdgfrb
          Gene Name:
          platelet derived growth factor receptor, beta polypeptide
          Gene Aliases:
          AI528809, CD140b, PDGFR-1, Pdgfr
          Chromosome Location:
          Chr.18: 61045150 - 61085068 on Build GRCm38
          UniGene ID:
          Mm.4146
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS207504
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pdgfrb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146268.1 NP_001139740.1 16 2668
          NM_008809.2 NP_032835.2 16 2665
          GenBank mRNA AK134785.1 - 17 2690
          AK143247.1 - 16 2648
          AK158145.1 - 10 1322
          BC055311.1 - 16 2680 30060666
          DQ979390.1 - 10 1321
          X04367.1 - 16 2374

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          kidney development
          tissue homeostasis
          glycosaminoglycan biosynthetic process
          protein phosphorylation
          nitrogen compound metabolic process
          chemotaxis
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          multicellular organism development
          positive regulation of cell proliferation
          positive regulation of phospholipase C activity
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of smooth muscle cell migration
          phosphorylation
          cell migration
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          adrenal gland development
          positive regulation of cell migration
          positive regulation of phosphoprotein phosphatase activity
          regulation of actin cytoskeleton organization
          positive regulation of collagen biosynthetic process
          cell migration involved in vasculogenesis
          intracellular signal transduction
          metanephric mesenchymal cell migration
          platelet-derived growth factor receptor-beta signaling pathway
          positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
          aorta morphogenesis
          cellular response to platelet-derived growth factor stimulus
          positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of mitotic nuclear division
          phosphatidylinositol metabolic process
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of smooth muscle cell proliferation
          skeletal system morphogenesis
          smooth muscle tissue development
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of chemotaxis
          cardiac myofibril assembly
          cell chemotaxis
          cardiac vascular smooth muscle cell differentiation
          cell migration involved in coronary angiogenesis
          retina vasculature development in camera-type eye
          positive regulation of ERK1 and ERK2 cascade
          smooth muscle cell chemotaxis
          metanephric glomerular mesangial cell proliferation involved in metanephros development
          metanephric glomerular capillary formation
          positive regulation of calcium ion import
          positive regulation of reactive oxygen species metabolic process
          positive regulation of DNA biosynthetic process
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          signal transducer activity
          platelet-derived growth factor-activated receptor activity
          platelet-derived growth factor beta-receptor activity
          receptor binding
          platelet-derived growth factor receptor binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          vascular endothelial growth factor binding
          phosphatidylinositol 3-kinase binding
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0