Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008810.2 | NP_032836.1 | 6 | 567 | ||
GenBank mRNA | AK145718.1 | - | 6 | 645 | ||
AK148459.1 | - | 6 | 588 | |||
AK167987.1 | - | 6 | 593 | |||
AK168152.1 | - | 6 | 617 | |||
AK169289.1 | - | 6 | 642 | |||
AK191895.1 | - | 2 | 94 | |||
BC007142.1 | - | 6 | 567 | 3600605 | ||
M76727.1 | - | 6 | 654 |