Search
Search

Pfkfb1| Gene Symbol: Pfkfb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008824.3 | NP_032850.1 | 8 | 847 | ||
| GenBank mRNA | AK132629.1 | - | 8 | 848 | ||
| BC132561.1 | - | 8 | 832 | 40130838 | ||
| BC132563.1 | - | 8 | 832 | 40130840 | ||