Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172695.2 | NP_766283.2 | 13 | 1900 | ||
GenBank mRNA | AK019070.1 | - | 13 | 1906 | ||
AK075934.1 | - | 13 | 1832 | |||
AK085027.1 | - | 3 | 1347 | |||
BC139355.1 | - | 13 | 1899 | 8862377 | ||
BC139356.1 | - | 13 | 1899 | 8862378 | ||
BC139773.1 | - | 13 | 1810 | 40111789 | ||
DV646766.1 | - | 5 | 386 |