Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011112.3 | NP_035242.1 | 16, 17 | 1736 | ||
GenBank mRNA | AK028940.1 | - | 16, 17 | 1734 | ||
AK090096.1 | - | 16, 17 | 1737 | |||
AK132704.1 | - | 16, 17 | 1723 | |||
BC060128.1 | - | 15, 16 | 1583 |