Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009099.2 | NP_033125.2 | 2 | 342 | ||
GenBank mRNA | AF220014.1 | - | 2 | 259 | ||
AF220015.1 | - | 2 | 259 | |||
AK089726.1 | - | 2 | 339 | |||
AK134144.1 | - | 2 | 311 | |||
AK137506.1 | - | 2 | 343 | |||
AK138249.1 | - | 2 | 345 | |||
BC005447.1 | - | 2 | 276 | 3493256 | ||
J03776.1 | - | 2 | 259 |