Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC9A1 SIRNA Assays ›
          Gene Symbol:
          Slc9a1
          Gene Name:
          solute carrier family 9 (sodium/hydrogen exchanger), member 1
          Gene Aliases:
          AW554487, Apnh, Mir5122, Nhe1, mir-5122, swe
          Chromosome Location:
          Chr.4: 133369772 - 133423702 on Build GRCm38
          UniGene ID:
          Mm.4312
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS209174
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc9a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016981.2 NP_058677.1 7, 8 2428
          GenBank mRNA AK150135.1 - 7, 8 2431
          AK154857.1 - 7, 8 2465
          BC051431.1 - 7, 8 1775 6529306
          BC052708.1 - 7, 8 2310 5702902
          U51112.1 - 7, 8 1971

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          transport
          ion transport
          cation transport
          sodium ion transport
          cellular sodium ion homeostasis
          regulation of pH
          embryo development
          response to acidic pH
          positive regulation of cardiac muscle hypertrophy
          regulation of cardiac muscle contraction by calcium ion signaling
          response to organic cyclic compound
          positive regulation of cell growth
          cellular response to insulin stimulus
          sodium ion transmembrane transport
          positive regulation of mitochondrial membrane permeability
          response to muscle stretch
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of action potential
          positive regulation of transcription from RNA polymerase II promoter
          regulation of intracellular pH
          positive regulation of NFAT protein import into nucleus
          regulation of sensory perception of pain
          cardiac muscle cell differentiation
          transmembrane transport
          positive regulation of calcineurin-NFAT signaling cascade
          neuron death
          cellular response to hypoxia
          cellular response to acidic pH
          cellular response to epinephrine stimulus
          regulation of the force of heart contraction by cardiac conduction
          sodium ion import across plasma membrane
          positive regulation of the force of heart contraction
          positive regulation of intracellular signal transduction
          hydrogen ion transmembrane transport
          positive regulation of calcium:sodium antiporter activity
          calmodulin binding
          antiporter activity
          solute:proton antiporter activity
          sodium:proton antiporter activity
          protein phosphatase 2B binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0