Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025287.2 | NP_079563.2 | 7 | 744 | ||
GenBank mRNA | AB071989.1 | - | 5 | 553 | ||
AK004669.1 | - | 7 | 714 | |||
AK028201.1 | - | 6 | 880 | |||
AK030746.1 | - | 6 | 931 | |||
AK132383.1 | - | 6 | 920 | |||
AK159482.1 | - | 6 | 897 | |||
AK160858.1 | - | 6 | 842 | |||
AK166581.1 | - | 6 | 854 | |||
AK168185.1 | - | 6 | 866 | |||
AK168546.1 | - | 6 | 933 | |||
AK189454.1 | - | 2 | 145 | |||
AY538613.1 | - | 5 | 556 | |||
BC043131.1 | - | 6 | 763 | 6811159 | ||
BC045205.1 | - | 7 | 714 | 4988521 | ||
BC064809.1 | - | 6 | 868 |