Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172136.1 | NP_001165607.1 | 7 | 873 | ||
NM_172456.3 | NP_766044.1 | 6 | 838 | |||
GenBank mRNA | AK028887.1 | - | 6 | 802 | ||
AK034654.1 | - | 7 | 835 | |||
AK051300.1 | - | 7 | 837 | |||
AK148426.1 | - | 6 | 819 | |||
AK162566.1 | - | 6 | 801 | |||
AK163555.1 | - | 6 | 802 | |||
AK184683.1 | - | 1 | 46 | |||
BC052171.1 | - | 5 | 644 | |||
BC059920.1 | - | 7 | 816 | |||
BC064730.1 | - | 7 | 888 | |||
BC094589.1 | - | 4 | 3324 | |||
BC119024.1 | - | 6 | 802 | 8733773 | ||
BC144729.1 | - | 7 | 837 |