Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145361.2 | NP_663336.2 | 5 | 1372 | ||
GenBank mRNA | AK046604.1 | - | 3 | 219 | ||
BC055704.1 | - | 4 | 439 | |||
BC083320.1 | - | 4 | 345 | |||
BC115682.1 | - | 4 | 432 | 40104217 | ||
BC115683.1 | - | 4 | 432 | 40104219 |