Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SULT1A1 SIRNA Assays ›
Gene Symbol:
Sult1a1
Gene Name:
sulfotransferase family 1A, phenol-preferring, member 1
Gene Aliases:
AI266890, PST, ST1A1, Stp, Stp1
Chromosome Location:
Chr.7: 126672865 - 126676483 on Build GRCm38
UniGene ID:
Mm.368982
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS209713
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sult1a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_133670.1
NP_598431.1
2
106
GenBank mRNA
AK002700.1
-
2
129
BC005413.1
-
2
106
3488459
L02331.1
-
2
122
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
catecholamine metabolic process
lipid metabolic process
xenobiotic metabolic process
steroid metabolic process
estrogen metabolic process
drug metabolic process
4-nitrophenol metabolic process
response to glucocorticoid
sulfation
nucleotide binding
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
aryl sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
tyrosine-ester sulfotransferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
protein homodimerization activity
3'-phosphoadenosine 5'-phosphosulfate binding
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-vsgnr:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0