Search
Search

Slc30a6| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252478.1 | NP_001239407.1 | 10 | 671 | ||
| NM_144798.6 | NP_659047.2 | 10 | 656 | |||
| GenBank mRNA | AF233346.1 | - | 9 | 608 | ||
| AF395840.1 | - | 9 | 598 | |||
| AK082907.1 | - | 10 | 633 | |||
| AK150342.1 | - | 10 | 632 | |||
| AK151120.1 | - | 10 | 632 | |||
| AK160903.1 | - | 10 | 660 | |||
| BC005753.1 | - | 2 | 119 | |||
| BC066162.1 | - | 10 | 623 | 30471027 | ||
| BC068169.1 | - | 10 | 645 | 30542277 | ||