Search
Search

Efhb| Gene Symbol: Efhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172497.3 | NP_766085.2 | 8, 9 | 1682 | ||
| GenBank mRNA | AK029550.1 | - | 8, 9 | 1683 | ||
| AK167235.1 | - | 5, 6 | 1310 | |||
| BC132103.1 | - | 8, 9 | 1673 | 40130380 | ||
| BC137880.1 | - | 8, 9 | 1673 | 8860899 | ||