Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145970.1 | NP_666082.2 | 16 | 1833 | ||
GenBank mRNA | BC016188.1 | - | 16 | 1833 | 4482793 | |
BC027028.1 | - | 15 | 1648 | 5363547 | ||
DQ329239.1 | - | 9 | 883 |