Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam149a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153535.2 | NP_705763.1 | 10 | 2406 | ||
GenBank mRNA | AK034229.1 | - | 10 | 1633 | ||
AK038552.1 | - | 10 | 1621 | |||
AK140440.1 | - | 10 | 2406 | |||
AK141607.1 | - | 10 | 2349 | |||
AK220518.1 | - | 10 | 1642 | |||
BC035537.1 | - | 10 | 2297 | 5369152 |