Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199467.2 | NP_955761.2 | 14 | 2538 | ||
GenBank mRNA | AK089531.1 | - | 7 | 1279 | ||
AK144799.1 | - | 13 | 2338 | |||
AK166072.1 | - | 14 | 2549 | |||
AY512923.1 | - | 9 | 1672 | |||
BC051674.1 | - | 5 | 985 | |||
BC094429.1 | - | 13 | 1882 | |||
BC137910.1 | - | 9 | 1448 | 8860929 | ||
BC145028.1 | - | 9 | 1448 | 9053576 |