Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172517.2 | NP_766105.2 | 5 | 571 | ||
GenBank mRNA | AK049247.1 | - | 5 | 558 | ||
AK154782.1 | - | 5 | 572 | |||
AK170520.1 | - | 5 | 569 | |||
AK171177.1 | - | 5 | 576 | |||
BC057632.1 | - | 6 | 444 | 4918318 |