Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177606.4 | NP_808274.2 | 13 | 2269 | ||
GenBank mRNA | AK029252.1 | - | 13 | 2271 | ||
AK173337.1 | - | 10 | 1924 | |||
BC125583.1 | - | 13 | 2234 | 40130098 | ||
BC137804.1 | - | 13 | 2231 | 8860823 |