Search
Search

Il1f9| Gene Symbol: Il1f9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153511.3 | NP_705731.2 | 4 | 553 | ||
| GenBank mRNA | AK081783.1 | - | 4 | 548 | ||
| AK156687.1 | - | 550 | ||||
| AY071843.1 | - | 3 | 279 | |||
| BC116227.1 | - | 3 | 279 | 40091521 | ||
| BC116228.1 | - | 3 | 279 | 40091522 | ||