Search
Search

Tes| Gene Symbol: Tes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207176.3 | NP_997059.1 | 6 | 1188 | ||
| GenBank mRNA | AK132249.1 | - | 6 | 1146 | ||
| AK145609.1 | - | 6 | 1137 | |||
| AK167101.1 | - | 6 | 1137 | |||
| AK167274.1 | - | 6 | 1145 | |||
| BC003808.1 | - | 8 | 1428 | 3589585 | ||
| BC010465.1 | - | 6 | 1144 | 3585670 | ||
| BC051103.1 | - | 6 | 1120 | |||
| CT010319.1 | - | 5 | 1023 | |||
| X78989.1 | - | 5 | 1017 | |||