Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR2E1 SIRNA Assays ›
          Gene Symbol:
          Nr2e1
          Gene Name:
          nuclear receptor subfamily 2, group E, member 1
          Gene Aliases:
          Mtl1, Mtll, TLL, Tlx, XTLL, fierce, frc, tailless
          Chromosome Location:
          Chr.10: 42561963 - 42583588 on Build GRCm38
          UniGene ID:
          Mm.287100
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS211941
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nr2e1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_152229.2 NP_689415.1 2 835
          GenBank mRNA AK132201.1 - 2 839
          BC056950.1 - 2 466
          BC057104.1 - 2 879 6401770
          S77482.1 - 2 878

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001113197, EU316216.

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          behavioral fear response
          aggressive behavior
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          brain development
          visual perception
          positive regulation of cell proliferation
          glial cell migration
          dentate gyrus development
          amygdala development
          olfactory bulb development
          layer formation in cerebral cortex
          forebrain generation of neurons
          cerebral cortex neuron differentiation
          anterior commissure morphogenesis
          cerebral cortex development
          extracellular matrix organization
          somatic stem cell population maintenance
          social behavior
          camera-type eye development
          negative regulation of apoptotic process
          steroid hormone mediated signaling pathway
          cell fate commitment
          negative regulation of neuron differentiation
          positive regulation of angiogenesis
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of astrocyte differentiation
          regulation of dendrite morphogenesis
          regulation of cellular component organization
          retina development in camera-type eye
          regulation of timing of neuron differentiation
          long-term synaptic potentiation
          regulation of cell migration involved in sprouting angiogenesis
          negative regulation of neural precursor cell proliferation
          positive regulation of neural precursor cell proliferation
          positive regulation of stem cell proliferation
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          protein binding
          zinc ion binding
          enzyme binding
          histone deacetylase binding
          sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4b6rj:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0