Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009433.3 | NP_033459.1 | 1 | 1015 | ||
GenBank mRNA | AF042180.1 | - | 1 | 979 | ||
AK053812.1 | - | 1 | 966 | |||
AK159432.1 | - | 1 | 996 | |||
AK166438.1 | - | 1 | 996 | |||
AK166805.1 | - | 1 | 944 | |||
AK166893.1 | - | 1 | 944 | |||
BC011213.1 | - | 1 | 962 | 4196841 |