Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136085.2 | NP_001129557.1 | 14 | 1670 | ||
NM_001276316.1 | NP_001263245.1 | 15 | 1762 | |||
NM_001276317.1 | NP_001263246.1 | 14 | 1665 | |||
NM_009457.4 | NP_033483.2 | 14 | 1665 | |||
GenBank mRNA | AK081808.1 | - | 14 | 1636 | ||
AK088057.1 | - | 14 | 1547 | |||
AK088528.1 | - | 14 | 1547 | |||
AK143416.1 | - | 14 | 1571 | |||
AK171667.1 | - | 14 | 1636 | |||
AK178731.1 | - | 2 | 102 | |||
AK191964.1 | - | 2 | 102 | |||
AK194651.1 | - | 2 | 102 | |||
AK195106.1 | - | 2 | 102 | |||
AK199948.1 | - | 2 | 102 | |||
AK200836.1 | - | 2 | 102 | |||
AK201099.1 | - | 2 | 102 | |||
AK201652.1 | - | 2 | 102 | |||
AK202271.1 | - | 2 | 102 | |||
AK204473.1 | - | 2 | 102 | |||
AK204546.1 | - | 2 | 102 | |||
AK205285.1 | - | 2 | 58 | |||
AK207789.1 | - | 2 | 102 | |||
AK213246.1 | - | 2 | 102 | |||
AK217264.1 | - | 2 | 102 | |||
AK217817.1 | - | 2 | 102 | |||
BC058630.1 | - | 14 | 1611 | 6402368 | ||
BC138200.1 | - | 14 | 1528 | 8861219 | ||
BC145984.1 | - | 14 | 1529 | 40131287 | ||
D10576.1 | - | 14 | 1531 |