Search
Search

Wars| Gene Symbol: Wars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164314.1 | NP_001157786.1 | 7, 8 | 1001 | ||
| NM_001164488.1 | NP_001157960.1 | 7, 8 | 934 | |||
| NM_011710.3 | NP_035840.3 | 7, 8 | 947 | |||
| GenBank mRNA | AK004541.1 | - | 7, 8 | 935 | ||
| AK007754.1 | - | 5, 6 | 563 | |||
| AK150090.1 | - | 8, 9 | 965 | |||
| AK150267.1 | - | 8, 9 | 1049 | |||
| AK152966.1 | - | 7, 8 | 965 | |||
| AK153337.1 | - | 7, 8 | 972 | |||
| BC003450.1 | - | 7, 8 | 933 | 2654231 | ||
| BC046232.1 | - | 7, 8 | 940 | 6813406 | ||
| CF731228.1 | - | 4, 5 | 453 | |||
| CT010258.1 | - | 6, 7 | 833 | |||