Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dzip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110017.1 | NP_001103487.1 | 16 | 2238 | ||
NM_027341.2 | NP_081617.1 | 15 | 1620 | |||
GenBank mRNA | AK039172.1 | - | 16 | 2216 | ||
BC052893.1 | - | 15 | 1428 | |||
BC117953.1 | - | 15 | 1443 | 40098141 | ||
BC117954.1 | - | 15 | 1449 | 40098143 | ||
CF536693.1 | - | 3 | 642 | |||
EU233997.1 | - | 8 | 1440 |