Search
Search

C330027C09Rik| Gene Symbol: C330027C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172616.2 | NP_766204.2 | 14, 15 | 1925 | ||
| GenBank mRNA | AK049356.1 | - | 14, 15 | 1927 | ||
| AK160639.1 | - | 14, 15 | 1912 | |||
| AK173190.1 | - | 13, 14 | 1663 | |||
| BC021874.1 | - | 3, 4 | 1120 | |||
| BC052662.1 | - | 5, 6 | 601 | |||
| BC137719.1 | - | 14, 15 | 1934 | 8860738 | ||