Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027060.1 | NP_081336.1 | 8 | 1607 | ||
NM_172618.2 | NP_766206.1 | 8 | 1607 | |||
GenBank mRNA | AK029578.1 | - | 8 | 1770 | ||
AK052664.1 | - | 8 | 1491 | |||
BC037031.1 | - | 6 | 1074 | |||
BC057897.1 | - | 8 | 1485 | 5031346 |