Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177796.1 | NP_001171267.1 | 12 | 1699 | ||
NM_001177797.1 | NP_001171268.1 | 11 | 1645 | |||
NM_146102.2 | NP_666214.1 | 13 | 1751 | |||
GenBank mRNA | AK048649.1 | - | 13 | 1405 | ||
AK087449.1 | - | 12 | 1335 | |||
AK220430.1 | - | 11 | 1268 | |||
BC031515.1 | - | 12 | 1301 | 3711746 |