Search
Search

Zfp207| Gene Symbol: Zfp207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130169.1 | NP_001123641.1 | 4, 5 | 653 | ||
| NM_001130170.1 | NP_001123642.1 | 4, 5 | 653 | |||
| NM_001130171.1 | NP_001123643.1 | 4, 5 | 653 | |||
| NM_011751.3 | NP_035881.1 | 4, 5 | 653 | |||
| GenBank mRNA | AB013357.1 | - | 4, 5 | 587 | ||
| AB032196.1 | - | 4, 5 | 534 | |||
| AK033497.1 | - | 4, 5 | 602 | |||
| AK087885.1 | - | 4, 5 | 653 | |||
| AK140139.1 | - | 4, 5 | 637 | |||
| AK146787.1 | - | 4, 5 | 616 | |||
| AK159683.1 | - | 4, 5 | 604 | |||
| BC003715.1 | - | 4, 5 | 594 | 3601357 | ||