Search
Search

Zfp207| Gene Symbol: Zfp207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130169.1 | NP_001123641.1 | 10 | 1188 | ||
| NM_001130170.1 | NP_001123642.1 | 9 | 1140 | |||
| NM_001130171.1 | NP_001123643.1 | 9 | 1095 | |||
| NM_011751.3 | NP_035881.1 | 9 | 1083 | |||
| GenBank mRNA | AB013357.1 | - | 9 | 1017 | ||
| AB032196.1 | - | 10 | 1069 | |||
| AK087885.1 | - | 9 | 1095 | |||
| AK140139.1 | - | 9 | 1124 | |||
| AK146787.1 | - | 10 | 1151 | |||
| AK159683.1 | - | 9 | 1046 | |||
| BC003715.1 | - | 9 | 1036 | 3601357 | ||