Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160038.1 | NP_001153510.1 | 8 | 954 | ||
NM_001160039.1 | NP_001153511.1 | 8 | 796 | |||
NM_001160040.1 | NP_001153512.1 | 7 | 850 | |||
NM_145518.2 | NP_663493.2 | 7 | 692 | |||
GenBank mRNA | AK034597.1 | - | 7 | 761 | ||
AK036926.1 | - | 7 | 557 | |||
AK078590.1 | - | 7 | 538 | |||
AK142711.1 | - | 7 | 572 | |||
AK146814.1 | - | 7 | 710 | |||
AK166604.1 | - | 8 | 643 | |||
AK167705.1 | - | 7 | 540 | |||
AK180817.1 | - | 5 | 343 | |||
AK213867.1 | - | 4 | 304 | |||
BC006660.1 | - | 7 | 579 | 3501043 | ||
BC015300.1 | - | 7 | 512 | 4236882 |