Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011769.4 | NP_035899.4 | 8 | 948 | ||
GenBank mRNA | AF111101.1 | - | 9 | 1071 | ||
AK086590.1 | - | 8 | 967 | |||
BC066196.1 | - | 8 | 948 | 30430385 |