Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144887.4 | NP_659136.1 | 11 | 2629 | ||
GenBank mRNA | AK028737.1 | - | 8 | 1195 | ||
AK082513.1 | - | 11 | 2554 | |||
AK145086.1 | - | 11 | 2556 | |||
AK173251.1 | - | 11 | 2584 | |||
AY894891.1 | - | 11 | 2621 | |||
BC020051.1 | - | 10 | 1599 | 3987696 | ||
BC027047.1 | - | 7 | 863 | |||
BC065155.1 | - | 11 | 2499 | 5706087 |