Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Madd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177719.1 | NP_001171190.1 | 27 | 4297 | ||
NM_001177720.1 | NP_001171191.1 | 25 | 3988 | |||
NM_145527.4 | NP_663502.3 | 26 | 4240 | |||
GenBank mRNA | AK122257.1 | - | 27 | 4139 | ||
AK163518.1 | - | 9 | 922 | |||
AY263980.1 | - | 27 | 4118 | |||
AY263981.1 | - | 25 | 4001 | |||
AY263982.1 | - | 24 | 3941 | |||
AY263983.1 | - | 25 | 3944 | |||
AY263984.1 | - | 24 | 3884 | |||
AY263985.1 | - | 24 | 3866 | |||
AY263986.1 | - | 24 | 3854 | |||
AY263987.1 | - | 24 | 3809 | |||
AY263988.1 | - | 23 | 3782 | |||
AY263989.1 | - | 23 | 3681 | |||
BC030369.1 | - | 3 | 223 | |||
BC042212.1 | - | 16 | 1978 | |||
BC063386.1 | - | 26 | 4041 | 5401819 |