Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039200.2 | NP_001034289.1 | 16 | 1956 | ||
NM_146157.4 | NP_666269.2 | 17 | 1965 | |||
GenBank mRNA | AF525925.1 | - | 7 | 804 | ||
AK049061.1 | - | 17 | 1956 | |||
AK077496.1 | - | 17 | 1956 | |||
AK089289.1 | - | 16 | 1935 | |||
AK129052.1 | - | 17 | 1918 | |||
AK138637.1 | - | 17 | 1938 | |||
AK170739.1 | - | 16 | 1935 | |||
BC048948.1 | - | 1 | 31 | |||
BC056952.1 | - | 12 | 1685 | |||
BC129804.1 | - | 17 | 1952 | 40087024 | ||
BC139366.1 | - | 17 | 1951 | 8862388 |