Search
Search

Cep104| Gene Symbol: Cep104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177673.2 | NP_808341.1 | 14 | 1969 | ||
| GenBank mRNA | AK080736.1 | - | 8 | 1259 | ||
| AK129163.1 | - | 13 | 1835 | |||
| AK143802.1 | - | 14 | 2385 | |||
| AK149826.1 | - | 14 | 2320 | |||
| BC046331.1 | - | 14 | 1969 | 6485859 | ||
| BC060106.1 | - | 7 | 1161 | |||