Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144918.2 | NP_659167.2 | 6 | 581 | ||
GenBank mRNA | AK158267.1 | - | 6 | 562 | ||
AK162926.1 | - | 6 | 505 | |||
BC016525.1 | - | 6 | 568 | 3601356 | ||
BU847995.1 | - | 6 | 584 |