Search
Search

Mybpc2| Gene Symbol: Mybpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146189.3 | NP_666301.2 | 22 | 2603 | ||
| GenBank mRNA | AK029282.1 | - | 22 | 2589 | ||
| AK132563.1 | - | 22 | 2593 | |||
| AK161146.1 | - | 22 | 2592 | |||
| BC029762.1 | - | 22 | 2573 | 3983351 | ||
| BC038869.1 | - | 7 | 866 | |||
| BC039184.1 | - | 22 | 2568 | 3980667 | ||