Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PICALM SIRNA Assays ›
          Gene Symbol:
          Picalm
          Gene Name:
          phosphatidylinositol binding clathrin assembly protein
          Gene Aliases:
          CALM, CLTH, PAP180, fit-1, fit1, mKIAA4114
          Chromosome Location:
          Chr.7: 90130220 - 90209447 on Build GRCm38
          UniGene ID:
          Mm.235175
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS214662
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Picalm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252520.1 NP_001239449.1 12 1530
          NM_001252521.1 NP_001239450.1 12 1530
          NM_001252522.1 NP_001239451.1 12 1530
          NM_001252523.1 NP_001239452.1 12 1530
          NM_001252524.1 NP_001239453.1 12 1530
          NM_146194.4 NP_666306.2 12 1530
          GenBank mRNA AK085472.2 - 9 764
          AK144960.1 - 12 1270
          AK162360.1 - 12 1223
          AK165066.1 - 7 642
          AK170373.1 - 12 1550
          AK220291.1 - 12 1246
          AY206701.1 - 12 1214
          BC011470.1 - 12 1240 2651109
          BC021491.1 - 12 1479 5345558
          BC023843.1 - 12 1278 5346041
          BC025566.1 - 12 1553 5322606
          BC027116.1 - 9 841
          BC057683.1 - 12 1481 3671962

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          receptor-mediated endocytosis
          multicellular organism development
          axonogenesis
          cell proliferation
          negative regulation of gene expression
          synaptic vesicle maturation
          vesicle-mediated transport
          endosomal transport
          hemopoiesis
          regulation of endocytosis
          receptor internalization
          regulation of protein localization
          cargo loading into vesicle
          positive regulation of transcription, DNA-templated
          negative regulation of receptor-mediated endocytosis
          clathrin coat assembly
          dendrite morphogenesis
          regulation of protein transport
          iron ion homeostasis
          clathrin-mediated endocytosis
          iron ion import into cell
          positive regulation of neuron death
          positive regulation of beta-amyloid formation
          regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          phospholipid binding
          1-phosphatidylinositol binding
          SH3 domain binding
          clathrin binding
          clathrin heavy chain binding
          clathrin adaptor activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0