Search
Search

Gdpd5| Gene Symbol: Gdpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201352.2 | NP_958740.2 | 15 | 2245 | ||
| GenBank mRNA | AK141189.1 | - | 15 | 2278 | ||
| AK154771.1 | - | 15 | 2382 | |||
| BC024955.1 | - | 14 | 1531 | 4953174 | ||
| BC026428.1 | - | 11 | 1178 | 4224574 | ||
| BC082585.1 | - | 15 | 2245 | 30608279 | ||
| BC119442.1 | - | 6 | 905 | |||
| BC119443.1 | - | 6 | 905 | |||