Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177977.1 | NP_001171448.1 | 2 | 361 | ||
NM_001177978.1 | NP_001171449.1 | 2 | 271 | |||
NM_146197.4 | NP_666309.1 | 2 | 271 | |||
GenBank mRNA | AK057650.1 | - | 2 | 271 | ||
AK140827.1 | - | 2 | 362 | |||
AK143757.1 | - | 2 | 267 | |||
AK165353.1 | - | 2 | 269 | |||
AK165401.1 | - | 2 | 260 | |||
BC031140.1 | - | 2 | 247 | 4972673 |