Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153507.2 | NP_705727.1 | 11 | 1177 | ||
GenBank mRNA | AK160238.1 | - | 11 | 1167 | ||
BC022128.1 | - | 11 | 1138 | 5097527 | ||
BC023348.1 | - | 6 | 475 | 5012573 | ||
BC031801.1 | - | 11 | 1177 | 3985959 |