Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177699.4 | NP_808367.2 | 9 | 1044 | ||
GenBank mRNA | AB041045.1 | - | 9 | 938 | ||
AK030737.1 | - | 9 | 1006 | |||
BC017144.1 | - | 8 | 1055 | |||
BC060654.1 | - | 9 | 1027 | 30354865 |