Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145607.3 | NP_663582.2 | 11 | 1377 | ||
GenBank mRNA | AK144842.1 | - | 11 | 1378 | ||
AK148102.1 | - | 13 | 1607 | |||
AK160894.1 | - | 13 | 1615 | |||
BC017545.1 | - | 5 | 591 | |||
BC072577.1 | - | 11 | 1370 | |||
BC079851.1 | - | 12 | 1432 | 6401930 |