Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172826.3 | NP_766414.3 | 4 | 1003 | ||
GenBank mRNA | AK041604.1 | - | 4 | 905 | ||
AK082856.1 | - | 4 | 920 | |||
AK141226.1 | - | 4 | 946 | |||
AY297430.2 | - | 4 | 1266 | |||
BC058740.1 | - | 4 | 931 | 6312377 |