Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022813.3 | NP_073724.1 | 6, 7 | 745 | ||
GenBank mRNA | AF295402.1 | - | 6, 7 | 749 | ||
AK161002.1 | - | 6, 7 | 745 | |||
AK171766.1 | - | 6, 7 | 736 | |||
BC003452.1 | - | 6, 7 | 719 | |||
BC014751.1 | - | 6, 7 | 722 | 4235556 |